Learn More
GA International Flame Resistant Permanent Marker for Metal Surfaces Black 1 each
Catalog No. NC1834384
Encompass_Preferred
Our flame-resistant permanent ink marker is intended for use on metal surfaces, including iron and stainless steel. With a wide working temperature range, especially for high-heat conditions (-196°C/-321°F to +1200°C/+2192°F), these markers are perfect for marking of areas to be cut, welded, brazed or for the identification of tools and equipment. Able to withstand direct flame contact, the markings will not burn off or fade, remaining detectable and legible in all weather conditions.
Catalog No. NC1834384 Supplier GA International Supplier No. FLM1B
Our flame-resistant permanent ink marker is intended for use on metal surfaces, including iron and stainless steel. With a wide working temperature range, especially for high-heat conditions (-196°C/-321°F to +1200°C/+2192°F), these markers are perfect for marking of areas to be cut, welded, brazed or for the identification of tools and equipment. Able to withstand direct flame contact, the markings will not burn off or fade, remaining detectable and legible in all weather conditions.