Learn More
Lakeland Industries Flame Resistant Coveralls
Catalog No. 502840536
Encompass_Preferred
Lakeland CAT 2 Inherently FR 7.0 oz. 88/12 Cotton coveralls are built tough and comfortable to protect the wearers from flash fires. C071 coveralls are configured with a quick release zipper, heavy duty snaps, pockets, and an elasticized waistband. Available in Navy and Khaki with reflective trim options, sizes SM-5X, this coverall is ideal when flash fire hazards are present.
Catalog No. 50-284-0536 Supplier Lakeland Industries Supplier No. C071RT13MD30
Lakeland CAT 2 Inherently FR 7.0 oz. 88/12 Cotton coveralls are built tough and comfortable to protect the wearers from flash fires. C071 coveralls are configured with a quick release zipper, heavy duty snaps, pockets, and an elasticized waistband. Available in Navy and Khaki with reflective trim options, sizes SM-5X, this coverall is ideal when flash fire hazards are present.