Learn More
National Safety Apparel™ Flame Resistant Nomex Fleece Cap
SDP
Catalog No. 17001698 Shop All National Safety Apparel Products
Encompass_Preferred
Change view
Click to view available options
Color: Navy
Catalog No.
17-001-698 Color: Navy
1 options
Catalog No. 17-001-698 Supplier National Safety Apparel™ Supplier No. H01FLCAP
For added warmth and FR protection, the flame resistant grid fleece cap offers a lightweight breathability option. With flat lock seams for maximum comfort, four-way stretch and high performance moisture wicking technology this cap will get your through cool weather temps easily.
  • Arc Rating: 24 cal/cm²
  • CAT 2
  • Fabric/Material: 8.5 oz. DuPont™ Nomex™ IIIA Fleece
  • Designed for comfort in cold winter temperatures
  • Excellent insulation
  • Moisture-wicking technology to keep you dryer
  • Breathable fabric
  • Made in the USA
Specifications
Size Medium
Material DuPont™ Nomex™ IIIA Fleece
Color Navy
Seam Type Stitched
Arc Flash Rating 24 cal/cm²
Certifications/Compliance CAT 2, ASTM F1506, OSHA 1910.269, NFPA 70 E, NFPA 2112, CSA Z462