Learn More
CritiCore Protective Wear Flame-Resistant Unisex Lab Coat
Catalog No. 18111316
Change view
Click to view available options
Size:
2X-Small
X-Small
Small
Medium
Large
X-Large
2X-Large
3X-Large
4X-Large
5X-Large
6X-Large
11 product options available for selection
Product selection table with 11 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
18-111-316 2X-Small
18-111-317 X-Small
18-111-318 Small
18-111-319 Medium
18-111-320 Large
18-111-321 X-Large
18-111-322 2X-Large
18-111-323 3X-Large
18-111-324 4X-Large
18-111-325 5X-Large
18-111-326 6X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 11 options available.
11 options
This item is not returnable. View return policy
Catalog No. 18-111-316 Supplier CritiCore Protective Wear Supplier No. 12L34940CXP63PXXSROY
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy
Designed to provide a superior level of protection against flash fire, electric arc (HRC 1), and other dangerous conditions. This Flame-Resistant lab coat has numerous applications in both laboratory and production settings.
  • Best in class flammability protection
  • Reusable Nomex™ FR fabric
  • Meets NFPA 70E and HRC 1 performance standards
  • Quick-release snap front
  • Safety sleeve cuff with tunneled elastic
  • FR identification patches on sleeves and back
  • Excellent heat-resistance properties
Specifications
Size 2X-Small
Gender Unisex
Material Nomex CXP6
Color Royal Blue
Closure Type Snaps
Cuff Style Elastic
Disposable Reusable
Sterility Non-sterile
No. of Pockets 3
Collar Type Lapel
Latex Free Yes
Arc Flash Rating HRC 1
Certifications/Compliance NFPA 2112 Certified Garment