Learn More
Perkin Elmer US LLC FIAS Sampling Probe Assembly for PerkinElmer AS-90/90A/90 Plus/91/93 Plus Autosampler
Catalog No. NC1776928
Encompass_Preferred
FIAS Standard Sampling Probe Assembly, 1.0 mm ID. Includes Stainless Steel Sampling Probe and Screw Fitting (B3000152) and FIAS Capillary Tubing Assembly (B0191060 – Replaces B0501044)
Catalog No. NC1776928 Supplier Perkin Elmer US LLC Supplier No. B3000161
Add to Cart
Add to Cart
FIAS Standard Sampling Probe Assembly, 1.0 mm ID. Includes Stainless Steel Sampling Probe and Screw Fitting (B3000152) and FIAS Capillary Tubing Assembly (B0191060 – Replaces B0501044)