Learn More
Uline Flammable Storage Cabinet, 45 Gallon, Yellow
Catalog No. NC0991593
Encompass
Yellow steel flammable liquid storage cabinet with self-closing doors, 45 gallon, 350lb shelf capacity, meets NFPA, IFC and OSHA standards, Yellow = for aerosols, gasoline and flammable liquids, doors automatically close if temperature reaches 165°F, 43 x 18 x 65""""
Catalog No. NC0991593 Supplier Uline Supplier No. H1564SY
Yellow steel flammable liquid storage cabinet with self-closing doors, 45 gallon, 350lb shelf capacity, meets NFPA, IFC and OSHA standards, Yellow = for aerosols, gasoline and flammable liquids, doors automatically close if temperature reaches 165°F, 43 x 18 x 65""""