Learn More
Uline Slimline Flammable Storage Cabinet - Manual Doors, Yellow, 12 Gallon
Supplier:  Uline H2218MY
Encompass
Ensure safety and provide storage for your flammable or combustible liquids.Use in narrow spaces.Double wall 18-gauge steel.350 lb. shelf capacity. Adjustable shelves.Sloped shelves safely direct spills away from containers.Recessed paddle handle, can be used with padlock.Continuous piano hinge provides smooth closure.Adjustable leveling feet for uneven surfaces.Meets NFPA, IFC and OSHA standards. FM approved.Yellow - For aerosols, gasoline and flammable liquids.
Catalog No. NC2866730