Learn More
Uline Undercounter Flammable Storage Cabinet - Manual Doors, Yellow, 22 Gallon
Catalog No. NC2865928
Encompass
Organize and safely store flammable liquids and aerosols in your auto or machine shop.Fits easily under workbenches with 35" clearance.Double wall 18-gauge steel.350 lb. shelf capacity. Adjustable shelves.Sloped shelves safely direct spills away from containers.Recessed paddle handle, can be used with padlock.Continuous piano hinge provides smooth closure.Adjustable leveling feet for uneven surfaces.Meets NFPA, IFC and OSHA standards. FM approved.Yellow - For aerosols, gasoline and flammable liquids.
Catalog No. NC2865928 Supplier Uline Supplier No. H4177MY
Organize and safely store flammable liquids and aerosols in your auto or machine shop.Fits easily under workbenches with 35" clearance.Double wall 18-gauge steel.350 lb. shelf capacity. Adjustable shelves.Sloped shelves safely direct spills away from containers.Recessed paddle handle, can be used with padlock.Continuous piano hinge provides smooth closure.Adjustable leveling feet for uneven surfaces.Meets NFPA, IFC and OSHA standards. FM approved.Yellow - For aerosols, gasoline and flammable liquids.