Learn More
HCL Labels Flammable Liquids - Cabinet or Secondary Containment Sign - 3.5" x 11" Adhesive Vinyl (Pack of 25)
SDP
Catalog No. NC1977718
Encompass_Preferred
Flammable Liquids Cabinet or Secondary Containment Label with GHS pictogram - Pack of 25 LabelsLABELS FOR CHEMICAL STORAGE CABINETS OR SECONDARY CONTAINER TRAYS•With GHS pictograms for quick hazard identification•Assists scientist with the proper segregation of chemicals•Printed using chemical resistant adhesive vinyl•Air egress adhesive that makes for an easy bubble-free application
Catalog No. NC1977718 Supplier HCL Labels Supplier No. SSLX210VN351125
Flammable Liquids Cabinet or Secondary Containment Label with GHS pictogram - Pack of 25 LabelsLABELS FOR CHEMICAL STORAGE CABINETS OR SECONDARY CONTAINER TRAYS•With GHS pictograms for quick hazard identification•Assists scientist with the proper segregation of chemicals•Printed using chemical resistant adhesive vinyl•Air egress adhesive that makes for an easy bubble-free application