Learn More
Cole-Parmer Vacuubrand 662511 Aluminum hose barb adapter, NW 16 to 10 mm
Catalog No. NC0283667 Shop All Cole-Parmer Products
Encompass
Adapters fit almost every kind of vacuum pump and accessory. Be sure to order clamps and co-seal or centering ring .
Catalog No. NC0283667 Supplier Cole-Parmer Supplier No. 3140050
Adapters fit almost every kind of vacuum pump and accessory. Be sure to order clamps and co-seal or centering ring .