Learn More
Cole-Parmer Vacuubrand 669041 Flange Adapter Vacuum Pump Fitting, Aluminum, NW 16 to NW 25
Catalog No. NC0283668 Shop All Cole-Parmer Products
Encompass
Adapters fit almost every kind of vacuum pump and accessory. Be sure to order clamps and co-seal or centering ring .
Catalog No. NC0283668 Supplier Cole-Parmer Supplier No. 3140010
Adapters fit almost every kind of vacuum pump and accessory. Be sure to order clamps and co-seal or centering ring .