Learn More
Ace Glass, Inc. Flange style vacuum trap 38mm OD x 280mm l, #40 clamp and O-ring
SDP
Catalog No. 50979862
Encompass_Preferred
38mm O.D. x 280mm long vacuum trap assembly with o-ring flange connection. Includes #40 stainless steel pinch clamp and Viton o-ring. This design has flexibility and ease of assembly/disassembly for cleaning.
Catalog No. 50-979-862 Supplier Ace Glass, Inc. Supplier No. 875510
38mm O.D. x 280mm long vacuum trap assembly with o-ring flange connection. Includes #40 stainless steel pinch clamp and Viton o-ring. This design has flexibility and ease of assembly/disassembly for cleaning.