Learn More
Ace Glass, Inc. Flange style vacuum trap 58mm OD x 280mm l, #75 clamp -ring
SDP
Catalog No. 50976369
Encompass_Preferred
58mm O.D. x 280mm long vacuum trap assembly with o-ring flange connection. Includes #75 stainless steel pinch clamp and Viton o-ring. This design has flexibility and ease of assembly/disassembly for cleaning.
Catalog No. 50-976-369 Supplier Ace Glass, Inc. Supplier No. 875517
58mm O.D. x 280mm long vacuum trap assembly with o-ring flange connection. Includes #75 stainless steel pinch clamp and Viton o-ring. This design has flexibility and ease of assembly/disassembly for cleaning.