Learn More
IDEX Health & Science Upchurch Flangeless Fitting | Short | Natural | PEEK | 1/4-28 Flat-Bottom | 1/8"""" OD | Natural 
Catalog No. NC1763495
Encompass_Preferred
Our Short Flangeless Fittings are designed to include both a nut and ferrule for your convenience. Flangeless fitting are dependable, easy to use and easy to replace. It has a 1/4-24 Flat-Bottom port configuration and is for 1/8"""" OD tubing.
Catalog No. NC1763495 Supplier IDEX Health & Science Upchurch Supplier No. XP335X
Our Short Flangeless Fittings are designed to include both a nut and ferrule for your convenience. Flangeless fitting are dependable, easy to use and easy to replace. It has a 1/4-24 Flat-Bottom port configuration and is for 1/8"""" OD tubing.