Learn More
Analytical Sales & Services Flangeless Fitting for 3/16" OD Tubing
Catalog No. 5000805897
Encompass
Large Bore Flangeless Fitting for 3/16" OD Tubing, 5/16-24 Flat-bottom port, 500 psi, PEEK/Tefzel (Nut and Ferrule)
Catalog No. 50-008-05897 Supplier Analytical Sales & Services Supplier No. 97132
Large Bore Flangeless Fitting for 3/16" OD Tubing, 5/16-24 Flat-bottom port, 500 psi, PEEK/Tefzel (Nut and Ferrule)