Learn More
Analytical Sales & Services Flangeless Fitting for 3/16" OD Tubing (10 pack)
Catalog No. 5000804995
Encompass
Large Bore Flangeless Fitting for 3/16" OD Tubing, 5/16-24 Flat-bottom port, 500 psi, PEEK/Tefzel (Nut and Ferrule)
Catalog No. 50-008-04995 Supplier Analytical Sales & Services Supplier No. 97132X
Add to Cart
Add to Cart
Large Bore Flangeless Fitting for 3/16" OD Tubing, 5/16-24 Flat-bottom port, 500 psi, PEEK/Tefzel (Nut and Ferrule)