Learn More
Cole-Parmer FLANGELESS FITTING, STANDARD K
Catalog No. NC2263612 Shop All Cole-Parmer Products
Encompass
Flangeless Fitting, Standard Knurl, Natural ETFE, 1/8" OD Tubing, 1/4-28 Flat-Bottom; 1/EA
This item is not returnable. View return policy
Catalog No. NC2263612 Supplier Cole-Parmer Supplier No. 0202094
Flangeless Fitting, Standard Knurl, Natural ETFE, 1/8" OD Tubing, 1/4-28 Flat-Bottom; 1/EA