Learn More
BIOTECH FLUIDICS LLC FLANGELESS FITTING SYSTEM
SDP
Catalog No. NC2158214
Encompass_Preferred
Flangeless Fitting System, Natural PEEK,Short, 1/4-28 for 1/16" OD Tubing (w/ P-235Nut and P-200 Ferrule), 10 Pack
This item is not returnable. View return policy
Catalog No. NC2158214 Supplier BIOTECH FLUIDICS LLC Supplier No. XP235X
Flangeless Fitting System, Natural PEEK,Short, 1/4-28 for 1/16" OD Tubing (w/ P-235Nut and P-200 Ferrule), 10 Pack