Learn More
Analytical Sales & Services Flangeless Nut for 1/16" OD Tubing
Catalog No. 5000806196
Encompass
Flangeless Nut for 1/16"OD Tubing, 10-32 Flat-bottom Ports PEEK/Tefzel
Catalog No. 50-008-06196 Supplier Analytical Sales & Services Supplier No. 95811X
Flangeless Nut for 1/16"OD Tubing, 10-32 Flat-bottom Ports PEEK/Tefzel