Learn More
IDEX Health & Science Upchurch FLANGELESS NUT EXTENDER TOOL
Catalog No. NC1919833
Encompass_Preferred
Idex Nut-Extender Tools, For Use With: Use to Tighten Any Standard Short or Extra Long Style Flangeless Nuts in Hard-to-Reach Places, Acetal, Nut-extender tool
Catalog No. NC1919833 Supplier IDEX Health & Science Upchurch Supplier No. P298
Idex Nut-Extender Tools, For Use With: Use to Tighten Any Standard Short or Extra Long Style Flangeless Nuts in Hard-to-Reach Places, Acetal, Nut-extender tool