Learn More
IDEX Health & Science Upchurch FLANGELESS NUT TEFZEL 1/8IN
Catalog No. NC2299836
Encompass_Preferred
Flangeless Nut Tefzel™ (ETFE), 1/4-28 Flat-Bottom, for 1/8" OD NaturalMinimum of 49 each per order.
Catalog No. NC2299836 Supplier IDEX Health & Science Upchurch Supplier No. P315
Flangeless Nut Tefzel™ (ETFE), 1/4-28 Flat-Bottom, for 1/8" OD NaturalMinimum of 49 each per order.