Learn More
Chemglass Life Sciences Nut, Flangeless, 1/8", Natural, Tefzel
SDP
Catalog No. CG1164N02
Encompass_Preferred
Nut, Flangeless, 1/8", Natural, Tefzel. Flangeless nut eliminates the need to flange tubing. Used with a flangeless ferrule for a leak-free connection of FEP tubing. Simply insert the tubing through the nut, attach the ferrule (flat end towards the PTFE fitting) to the end of the tubing and finger tighten the nut into place. Nut has a 1/4in- 28 thread, and is molded out of natural Tefzel, a soft fluoropolymer resistant to chemical attack.
Catalog No. CG1164N02 Supplier Chemglass Life Sciences Supplier No. CG1164N02
Nut, Flangeless, 1/8", Natural, Tefzel. Flangeless nut eliminates the need to flange tubing. Used with a flangeless ferrule for a leak-free connection of FEP tubing. Simply insert the tubing through the nut, attach the ferrule (flat end towards the PTFE fitting) to the end of the tubing and finger tighten the nut into place. Nut has a 1/4in- 28 thread, and is molded out of natural Tefzel, a soft fluoropolymer resistant to chemical attack.