Learn More
Hach Company Flare, Cell/Waste - 13 cm (Replacement for FIA cell/waste lines)
Catalog No. NC2100323 Shop All Hach Company Products
Encompass
This 13 cm flare is a replacement component for cell/waste lines used in flow injection analysis (FIA) systems.
  • 13 cm length.
  • Replacement part for cell/waste lines in flow injection analysis systems.
  • Functions as a flare to direct waste away from the measurement cell.
Catalog No. NC2100323 Supplier Hach Company Supplier No. 50065L
This 13 cm flare is a replacement component for cell/waste lines used in flow injection analysis (FIA) systems.
  • 13 cm length.
  • Replacement part for cell/waste lines in flow injection analysis systems.
  • Functions as a flare to direct waste away from the measurement cell.