Learn More
Hach Company Hach Flare Flow Cell Carrier Line, 35 cm - replacement carrier line for Lachat flow cell assemblies
Catalog No. NC2157413 Shop All Hach Company Products
Encompass
The Hach Flare Flow Cell Carrier Line (35 cm) is a replacement carrier line used with flow cell assemblies in flow injection analysis equipment. It provides the specified 35 cm length carrier line for positioning a flow cell within the analysis path.
  • 35 cm length.
  • Flow cell carrier line for flow cell assemblies.
  • Replacement part for flow injection analysis systems.
  • Provides a consistent carrier length for repeatable positioning of the flow cell.
This item is not returnable. View return policy
Catalog No. NC2157413 Supplier Hach Company Supplier No. 50067L
The Hach Flare Flow Cell Carrier Line (35 cm) is a replacement carrier line used with flow cell assemblies in flow injection analysis equipment. It provides the specified 35 cm length carrier line for positioning a flow cell within the analysis path.
  • 35 cm length.
  • Flow cell carrier line for flow cell assemblies.
  • Replacement part for flow injection analysis systems.
  • Provides a consistent carrier length for repeatable positioning of the flow cell.