Learn More
Grainger Proto Combination Flare Nut Wrench Set, Alloy Steel, 6-Point, 3/8 in. to 3/4 in., 7-Piece Set
Catalog No. 502793525
Encompass_Preferred
This 7-piece, 6-point flare nut combination wrench set is designed for professional use, offering precision and durability for various mechanical applications. Constructed from high-quality alloy steel with a corrosion-resistant satin finish, these tools are built to withstand demanding industrial environments.
  • 6-point flare nut opening type with TorquePlus™ system for improved grip and reduced fastener rounding
  • Satin finish provides corrosion resistance and a slip-resistant grip
  • Includes seven wrenches ranging from 3/8 in. to 3/4 in.
  • Offset head design with a 15° head offset angle for better clearance in confined spaces
  • Includes a red wrench roll for organized storage and portability
  • Compliant with ASME B107.100 standards
  • Tether capable for increased safety in elevated work environments
Catalog No. 50-279-3525 Supplier Grainger Supplier No. 1AKT8
This 7-piece, 6-point flare nut combination wrench set is designed for professional use, offering precision and durability for various mechanical applications. Constructed from high-quality alloy steel with a corrosion-resistant satin finish, these tools are built to withstand demanding industrial environments.
  • 6-point flare nut opening type with TorquePlus™ system for improved grip and reduced fastener rounding
  • Satin finish provides corrosion resistance and a slip-resistant grip
  • Includes seven wrenches ranging from 3/8 in. to 3/4 in.
  • Offset head design with a 15° head offset angle for better clearance in confined spaces
  • Includes a red wrench roll for organized storage and portability
  • Compliant with ASME B107.100 standards
  • Tether capable for increased safety in elevated work environments