Learn More
R&D Systems™ FLARE™ Slides

Description
- The CometAssay, or single cell gel electrophoresis assay, permits the quantification of DNA damage in individual cells based on the migration of DNA structures in an electric field
- As a result of the migration, Comet tail shaped images are formed and the amount of DNA in the Comet tail as well as the distance of migration, can be a measure of damage
- FLARE™ Slides are convenient, simple, and reliable format for sample preparation, processing, analysis, and storage for the CometAssay and FLARE™ Assay
Specifications
Specifications
| Content And Storage | Store the unopened product at room temperature. Do not use past expiration date. |
| Description | Sample-ready pre-treated glass slides |
| Format | Hydrophobic Barrier, Specially-treated Surface, Writing Surface |
| For Use With (Equipment) | For use with CometAssay™ Kit and FLARE™ Assay Kits, reagents, and protocols |
| For Use With (Application) | Sample preparation, processing, analysis, and storage for the CometAssay™ and FLARE™ Assay |
| Product Line | R&D Systems |
| Product Type | Slide |
| Quantity | 100 Slides |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.