Learn More
Glass Expansion Contour Flared End Solva Flex Pump Tube 2tag, 140mm spacing, 1.42mm ID, Yellow/Yellow (PKT 6)
SDP
Supplier:  Glass Expansion 1.42YLWSFF
Encompass_Preferred
Contour Flared End Solva Flex Pump Tube 2tag, 140mm spacing, 1.42mm ID Yellow/Yellow (PKT 6).Tag Colors: Yellow/YellowID mm: 1.42Wall Thickness mm: 0.86OD mm: 3.14Overall Length: 455mm (140mm spacing between tags)Number of Tubes Per Pack: 6Physical Properties: Thermoplastic, PVC based material with plasticizerMaterial: Made from Tygon 4040 or equivalent.Special Properties: Special tubing for hydrocarbons, petroleum products and distillates.Chemical Compatibility: Ideal for gasoline, kerosene, heating oils, cutting liquids and coolants based on glycols.Limitations: Not recommended for strong acids and alkalis, foodstuffs, beverages and medicines. Potential leaching of plasticizersGlass Expansion is ISO 9001:2015 certified by AB Certification, No. AU4501-9001
Catalog No. 50-190-6867