Learn More
Glass Expansion Contour Flared End Viton Pump Tube 2tag, 127mm spacing, 1.30mm ID, Grey/Grey (PKT 6)
SDP
Supplier:  Glass Expansion VS1.30GRYF
Encompass_Preferred
Contour Flared End Viton Pump Tube 2tag, 127mm spacing, 1.30mm ID Gray/Gray (PKT 6).Tag Colors: Gray/GrayID mm: 1.30Wall Thickness mm: 0.90OD mm 3.10Overall Length: 175mm (127mm spacing between tags)Number of Tubes Per Pack: 6Physical Properties: Fluoropolymer elastomerSpecial Properties: Special tubing for concentrated acids and corrosive solventsMaterial: Made from Fluran HC F-5500-A or equivalent.Limitations: Limited service lifeGlass Expansion is ISO 9001:2015 certified by AB Certification, No. AU4501-9001
Catalog No. 50-190-7740