Learn More
Glass Expansion Contour Flared End Viton Pump Tube 2tag, 127mm spacing, 1.65mm ID, Blue/Blue (PKT 6)
SDP
Supplier:  Glass Expansion VS1.65BLUF
Encompass_Preferred
Contour Flared End Viton Pump Tube 2tag, 127mm spacing, 1.65mm ID Blue/Blue (PKT 6).Tag Colors: Blue/BlueID mm: 1.65Wall Thickness mm: 0.90OD mm 3.45Overall Length: 175mm (127mm spacing between tags)Number of Tubes Per Pack: 6Physical Properties: Fluoropolymer elastomerSpecial Properties: Special tubing for concentrated acids and corrosive solventsMaterial: Made from Fluran HC F-5500-A or equivalent.Limitations: Limited service lifeGlass Expansion is ISO 9001:2015 certified by AB Certification, No. AU4501-9001
Catalog No. 50-190-7746