Learn More
Perkin Elmer US LLC Flared PVC MP2 Two-Stop Peristaltic Pump Tubing - 0.27 mm I.D., Orange/Blue, Pkg. 12
Catalog No. 502105037
Encompass_Preferred
MPP PVC Tubing Flared, 0.27mm, orange blue, 12/pk. Flared tubing is easier to change and more reliably connected than non-flared small-bore tubing.
Catalog No. 50-210-5037 Supplier Perkin Elmer US LLC Supplier No. N8145196
MPP PVC Tubing Flared, 0.27mm, orange blue, 12/pk. Flared tubing is easier to change and more reliably connected than non-flared small-bore tubing.