Learn More
Perkin Elmer US LLC Flared PVC MP2 Two-Stop Peristaltic Pump Tubing - 0.64 mm I.D., Orange/White, Pkg. 12
Catalog No. 502105039
Encompass_Preferred
MPP PVC Tubing Flared, 0. 64mm, orange white, 12/pk. Flared tubing is easier to change and more reliably connected than non-flared small-bore tubing.
Catalog No. 50-210-5039 Supplier Perkin Elmer US LLC Supplier No. N8145201
MPP PVC Tubing Flared, 0. 64mm, orange white, 12/pk. Flared tubing is easier to change and more reliably connected than non-flared small-bore tubing.