Learn More
Perkin Elmer US LLC Flared PVC Two-Stop Peristaltic Pump Tubing - 0.64 mm I.D., Orange/White, Pkg. 12
Catalog No. 502112186
Encompass_Preferred
This flared PVC Two-Stop Pump Tubing is for use with the standard pump in Optima, Avio, and NexION 300/350 series instruments. This tubing ships as a package of 12.
Catalog No. 50-211-2186 Supplier Perkin Elmer US LLC Supplier No. N0777711
This flared PVC Two-Stop Pump Tubing is for use with the standard pump in Optima, Avio, and NexION 300/350 series instruments. This tubing ships as a package of 12.