Learn More
Glass Expansion Peristlatic Pump Tubing, Solva, 2-tag, Glass Expansion. For large pump, Orange/White, 0.64 mm ID, with flared ends. For hydrocarbons, petroleum products and distillates. Pack of 6
SDP
Supplier:  Glass Expansion 0.64OWSFF
Encompass_Preferred
Contour Flared End Solva Flex Pump Tube 2tag, 140mm spacing, 0.64mm ID Orange/White (PKT 6).Tag Colors: Orange/WhiteID mm: 0.64Wall Thickness mm: 0.91OD mm: 2.46Overall Length: 455mm (140mm spacing between tags)Number of Tubes Per Pack: 6Physical Properties: Thermoplastic, PVC based material with plasticizerMaterial: Made from Tygon 4040 or equivalent.Special Properties: Special tubing for hydrocarbons, petroleum products and distillates.Chemical Compatibility: Ideal for gasoline, kerosene, heating oils, cutting liquids and coolants based on glycols.Limitations: Not recommended for strong acids and alkalis, foodstuffs, beverages and medicines. Potential leaching of plasticizersGlass Expansion is ISO 9001:2015 certified by AB Certification, No. AU4501-9001
Catalog No. 50-169-4697