Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phenomenex Inc Flash 20m C18 100

Catalog No. 502529271 Shop All Phenomenex Products
Encompass

Sorbent, 5000g/Vial Spherical

Catalog No. 50-252-9271 Supplier Phenomenex Inc Supplier No. FSO12050000
May include imposed supplier surcharges.
Add to Cart
Add to Cart
Request a Quote

missing translation for 'validMainframeMsg'

Sorbent, 5000g/Vial Spherical

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.