Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phenomenex Inc Sorbent, 1000g/Vial Spherical

Catalog No. 502395294 Shop All Phenomenex Products
Encompass

Flash 20m Silica 100 FSS1201000-0

Catalog No. 50-239-5294 Supplier Phenomenex Inc Supplier No. FSS12010000
May include imposed supplier surcharges.
Add to Cart
Add to Cart
Request a Quote

missing translation for 'validMainframeMsg'

Flash 20m Silica 100 FSS1201000-0

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.