Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phenomenex Inc Sorbent, 25kg/pk Reagent Grade

Catalog No. 502394827 Shop All Phenomenex Products
Encompass

Flash 50m Silica 60 SS605025-P

Catalog No. 50-239-4827 Supplier Phenomenex Inc Supplier No. SS605025P
May include imposed supplier surcharges.
Add to Cart
Add to Cart
Request a Quote

missing translation for 'validMainframeMsg'

Flash 50m Silica 60 SS605025-P

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.