Learn More
MilliporeSigma™ Supelco™ Flash Arrestor
Catalog No. 111009144 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Max. Pressure:
50 psig (3.5 kg/cm2)
Catalog No. Max. Pressure
11-100-9144 50 psig (3.5 kg/cm2)
1 options
Catalog No. 11-100-9144 Supplier MilliporeSigma™ Supelco™ Supplier No. 23315
Add to Cart
Edge
Add to Cart
Install a flash arrestor downstream of a hydrogen gas cylinder or a hydrogen generator.
In event of a flashback, the flash arrestor diverts the flame into three feet (1 m) of tubing, where the flame is extinguished and the heat is absorbed. The shock wave preceding the flashback closes and locks the arrestor's shutoff valve, eliminating the flow of gas. Inlet/outlet fittings are 1/4 in. female NPT. For use to 50 psig (3.5 kg/cm2). Meets Occupational Safety and Health Administration (OSHA) and National Fire Protection Agency (NFPA) codes. Factory Mutual approved.
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Application) Flash Chromatography
Max. Pressure 50 psig (3.5 kg/cm2)