Learn More
Phenomenex Inc Flash C18 sorbent
Catalog No. 502397707 Shop All Phenomenex Products
Encompass
Flash C18 sorbent 50-75μm; 100A; 5000g/bottle; Amorphous
Catalog No. 50-239-7707 Supplier Phenomenex Inc Supplier No. FCO24050000
Add to Cart
Add to Cart
Request a Quote
Flash C18 sorbent 50-75μm; 100A; 5000g/bottle; Amorphous