Learn More
WELCH MATERIALS INC Welch MaterialsTM WelFlashTM Flash Cartridge, SiO2-Ⅱ, Irregular Silica, 80g, 10/pk
Catalog No. 501963639
Encompass_Preferred
Flash Cartridge, irregular silica, 40-63μm, normal phase, size: 80g, pore size: 60Å, luer lock connections, PP tube, max. pressure: 14 bar, 10/pk, compatible with most FPLC system.
Catalog No. 50-196-3639 Supplier WELCH MATERIALS INC Supplier No. 00101050532
Flash Cartridge, irregular silica, 40-63μm, normal phase, size: 80g, pore size: 60Å, luer lock connections, PP tube, max. pressure: 14 bar, 10/pk, compatible with most FPLC system.