Learn More
Sorbent Technologies Inc Flash Cartridge, Screw Top, Luer Lock end fittings, Includes top and bottom frit, o-ring, dispersing insert, PP, 40g, 72ml, 12/pk
SDP
Catalog No. NC0946457
Encompass_Preferred
These empty flash cartridges come ready for you to easily pack and repack them with any adsorbent. This solvent resistant polypropylene cartridge has a volume of 53ml. Cartridges are rated at 200 psi (13.8 bar). The cartridge has a luer lock connection at the top and bottom. Pack includes cartridges, top and bottom frits, o-rings, and dispersing inserts. For use with Sorbent Technologies' Silica Gels, Aluminas, and other adsorbents.
Catalog No. NC0946457 Supplier Sorbent Technologies Inc Supplier No. FCSTLL4012
These empty flash cartridges come ready for you to easily pack and repack them with any adsorbent. This solvent resistant polypropylene cartridge has a volume of 53ml. Cartridges are rated at 200 psi (13.8 bar). The cartridge has a luer lock connection at the top and bottom. Pack includes cartridges, top and bottom frits, o-rings, and dispersing inserts. For use with Sorbent Technologies' Silica Gels, Aluminas, and other adsorbents.