Learn More
LUKNOVA INC Empty Column, 330 Ml Volume To Fit 240 Gram Silica, Luer Inlet, Polypropylene Structure, Polyethylene Frit Pore Size 20 µm, Chamber Ø 65 X 200 Mm. 4 Per Pack.
Catalog No. 50918475
Encompass
Luknova Patented Supersep Empty Flash Column, 330 Ml Volume Able To Fit 240 Gram Of Standard Silica (230-400 Mesh), Luerlock Inlet, Polypropylene Structure, Polyethylene Frit With Pore Size 20 µm, Empty Chamber Size Ø 65 X 200 Mm. These Columns Are Disposable And Each Unit Comes At 4 Columns Per Pack . They Are Designed To Be Assembled Easily And Fit Directly In The Teledyne Isco, Biotage Isolera, Interchim Puriflash, Yamazen, Santai, Agela, And Other Flash Systems. Great To Pack Your Own Columns With Youe Own Or Special Packing Materials. More Top And Bottom Frits Are Available.
Catalog No. 50-918-475 Supplier LUKNOVA INC Supplier No. FC0032401
Luknova Patented Supersep Empty Flash Column, 330 Ml Volume Able To Fit 240 Gram Of Standard Silica (230-400 Mesh), Luerlock Inlet, Polypropylene Structure, Polyethylene Frit With Pore Size 20 µm, Empty Chamber Size Ø 65 X 200 Mm. These Columns Are Disposable And Each Unit Comes At 4 Columns Per Pack . They Are Designed To Be Assembled Easily And Fit Directly In The Teledyne Isco, Biotage Isolera, Interchim Puriflash, Yamazen, Santai, Agela, And Other Flash Systems. Great To Pack Your Own Columns With Youe Own Or Special Packing Materials. More Top And Bottom Frits Are Available.