Learn More
Medchemexpress LLC FlAsH-EDT2 | 212118-77-9 | MFCD27977924 | ≥99.0% | 664.50 | C24H18As2O5S4 | 5 MG
SDP
Catalog No. 5000262871
Encompass_Preferred
FlAsH-EDT2 is a membrane-permeant biarsenical fluorescent probe for selective labeling of proteins containing tetracysteine (Cys4) motifs. It binds tetracysteine sequences with high affinity and becomes highly fluorescent upon binding, making it suitable for live-cell imaging, protein localization, and tracking studies.
  • Membrane-permeant probe for live-cell labeling.
  • Binds tetracysteine (Cys4) motifs with high affinity.
  • Fluoresces strongly upon binding, enabling sensitive detection.
  • Compatible with fluorescence microscopy and protein tracking.
  • Available in small milligram pack sizes for research use.
  • High purity suitable for biological assays.
Catalog No. 50-002-62871 Supplier Medchemexpress LLC Supplier No. HY1304125MG
FlAsH-EDT2 is a membrane-permeant biarsenical fluorescent probe for selective labeling of proteins containing tetracysteine (Cys4) motifs. It binds tetracysteine sequences with high affinity and becomes highly fluorescent upon binding, making it suitable for live-cell imaging, protein localization, and tracking studies.
  • Membrane-permeant probe for live-cell labeling.
  • Binds tetracysteine (Cys4) motifs with high affinity.
  • Fluoresces strongly upon binding, enabling sensitive detection.
  • Compatible with fluorescence microscopy and protein tracking.
  • Available in small milligram pack sizes for research use.
  • High purity suitable for biological assays.