Learn More
AccuStandard, Inc FLASH POINT STANDARD 250ML
SDP
Catalog No. NC9290291
Encompass
COC D-92 - Nominal Flash Point (230 ?C). 250 mL. 8012-95-1
Catalog No. NC9290291 Supplier AccuStandard, Inc Supplier No. ASTMP13204
COC D-92 - Nominal Flash Point (230 ?C). 250 mL. 8012-95-1