Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Flash Point Standard COC ASTM D 92 (85.0°C), MilliporeSigma™ Supelco™

Catalog No. 111002466 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
3 x 80 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-100-2466 3 x 80 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-100-2466 Supplier MilliporeSigma Supelco Supplier No. CRMUCOCLOW3X80ML
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Certified reference material designed for flash point calibration.

TRUSTED_SUSTAINABILITY

Specifications

CAS 64742-47-8
Type Organic Standard
Quantity 3 x 80 mL
Grade Primary Reference Standard
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.