Learn More
Phenomenex Inc Flash Spherical NH2
Catalog No. 502399892 Shop All Phenomenex Products
Encompass_Preferred
Flash Spherical NH2 Claricep Flash columns, spherical NH2 20-35µm, 100A, 40g/Cartridge
Catalog No. 50-239-9892 Supplier Phenomenex Inc Supplier No. SN2300400
Flash Spherical NH2 Claricep Flash columns, spherical NH2 20-35µm, 100A, 40g/Cartridge