Learn More
Phenomenex Inc Flash Spherical Silica
Catalog No. 502412845 Shop All Phenomenex Products
Encompass_Preferred
Flash Spherical Silica Spherical; 20-35μm; 60A; 20g/branch
Catalog No. 50-241-2845 Supplier Phenomenex Inc Supplier No. SS130020P
Flash Spherical Silica Spherical; 20-35μm; 60A; 20g/branch