Learn More
Phenomenex Inc Flash Spherical Silica sorbent
Catalog No. 502398356 Shop All Phenomenex Products
Encompass
Flash Spherical Silica sorbent 20-35μm; 100A; 5000g/bottle
Catalog No. 50-239-8356 Supplier Phenomenex Inc Supplier No. FSS23050000
Add to Cart
Add to Cart
Request a Quote
Flash Spherical Silica sorbent 20-35μm; 100A; 5000g/bottle