Learn More
Phenomenex Inc Flash Spherical Silica(CM)
Catalog No. 502397440 Shop All Phenomenex Products
Encompass_Preferred
Flash Spherical Silica(CM) Spherical; 20-35μm; 100A; 20g/branch
Catalog No. 50-239-7440 Supplier Phenomenex Inc Supplier No. SM2300200
Flash Spherical Silica(CM) Spherical; 20-35μm; 100A; 20g/branch