Learn More
Lonza Walkersville FlashGel DNA Cassettes
Catalog No. NC0174862
Encompass_Preferred
FlashGel DNA Cassettes are essential components of the FlashGel System for fast electrophoresis of DNA. Double tier FlashGel DNA Cassettes are the ideal sample screening tool for checking up to 32 PCR or restriction fragment samples quickly without having to plan your day around agarose gels or to verify your DNA sample integrity prior to downstream analysis. The FlashGel System is the fastest way to separate DNA and the best way to watch DNA migration as it happens.Benefits:5 minute separation and recoverySee bands in as little as 2 minutesIdeal for small (< 1 kb) DNA fragmentsTest up to 32 samplesQuick checks for enzymatic reactions (e.g. PCR amplification, T7 endouclease cleavage)Real-time separation and documentationWatch band migration as it happensPhotograph gels at the bench without DNA damaging UV lightOutstanding sensitivity and resolutionApplications:DNA Electrophoresis Content:FlashGel DNA Cassettes 2.2% 16+1 double-tier 9 pack
Catalog No. NC0174862 Supplier Lonza Walkersville Supplier No. 57032
FlashGel DNA Cassettes are essential components of the FlashGel System for fast electrophoresis of DNA. Double tier FlashGel DNA Cassettes are the ideal sample screening tool for checking up to 32 PCR or restriction fragment samples quickly without having to plan your day around agarose gels or to verify your DNA sample integrity prior to downstream analysis. The FlashGel System is the fastest way to separate DNA and the best way to watch DNA migration as it happens.Benefits:5 minute separation and recoverySee bands in as little as 2 minutesIdeal for small (< 1 kb) DNA fragmentsTest up to 32 samplesQuick checks for enzymatic reactions (e.g. PCR amplification, T7 endouclease cleavage)Real-time separation and documentationWatch band migration as it happensPhotograph gels at the bench without DNA damaging UV lightOutstanding sensitivity and resolutionApplications:DNA Electrophoresis Content:FlashGel DNA Cassettes 2.2% 16+1 double-tier 9 pack