Learn More
Lonza Walkersville FlashGel DNA Starter Kit
Catalog No. 502120221
Encompass
The FlashGel DNA Starter Kit allows researchers to quickly begin using FlashGel DNA Cassettes for fast electrophoresis of DNA. The FlashGel DNA Starter kit contains all the elements needed to immediately begin testing DNA samples. FlashGel DNA Cassettes are the ideal sample screening tools to check up to 32 PCR or restriction fragments quickly without having to plan your day around agarose gels. Monitor gel runs right at the bench, without UV light. The highly sensitive system allows a 5-20X reduction in DNA levels - saving both time and money.Benefits:5 minute separation and recovery, see bands in as little as 2 minutesRecover samples directly without UV light band excision or purificationReal-time separation and documentation, watch band migration as it happensPhotograph gels at the bench without DNA damaging UV lightOutstanding sensitivity and resolutionContent:FlashGel DockFlashGel DNA CassettesFlashGel Loading DyeFlashGel Marker 100 - 4000bp
Catalog No. 50-212-0221 Supplier Lonza Walkersville Supplier No. 57026
The FlashGel DNA Starter Kit allows researchers to quickly begin using FlashGel DNA Cassettes for fast electrophoresis of DNA. The FlashGel DNA Starter kit contains all the elements needed to immediately begin testing DNA samples. FlashGel DNA Cassettes are the ideal sample screening tools to check up to 32 PCR or restriction fragments quickly without having to plan your day around agarose gels. Monitor gel runs right at the bench, without UV light. The highly sensitive system allows a 5-20X reduction in DNA levels - saving both time and money.Benefits:5 minute separation and recovery, see bands in as little as 2 minutesRecover samples directly without UV light band excision or purificationReal-time separation and documentation, watch band migration as it happensPhotograph gels at the bench without DNA damaging UV lightOutstanding sensitivity and resolutionContent:FlashGel DockFlashGel DNA CassettesFlashGel Loading DyeFlashGel Marker 100 - 4000bp